Tisk…
Folic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9
M-Clarity™ quality level = MQ200
F8798-100G F8798-25G F8798-5G
SIALF8798-100GEA 12870 CZK
SIALF8798-100G SIALF8798-25G SIALF8798-5G
Folic acid, Sigma-Aldrich®
Folic acid
Vzorec: C₁₉H₁₉N₇O₆
MW: 441.41 g/mol
MDL Number: MFCD00079305
CAS číslo: 59-30-3
EINECS: 200-419-0