Tisk…
Folic acid
Dodavatel: PanReac AppliChem
Synonyms: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9
A7092.0100 A7092.0025
APPCA7092.0100EA 9420 CZK
APPCA7092.0100 A7092.0025
Folic acid
Folic acid
Vzorec: C₁₉H₁₉N₇O₆
MW: 441,41 g/mol
MDL Number: MFCD00079305
CAS číslo: 59-30-3
EINECS: 200-419-0