N(α)-Boc-L-histidine ≥98%
Dodavatel: Thermo Scientific
Synonyms:
Boc-His-OH
B22042.03
B22042.06
B22042.03EA
749
CZK
B22042.03
B22042.06
N(α)-Boc-L-histidine ≥98%
N(α)-Boc-L-histidine
Purity: 98+%
CAS No.: 17791-52-5
Molecular Formula: C11H17N3O4
Formula Weight: 255.27
Melting Point: ca 198° dec.
Beilstein Reference: 755289
Vzorec:
C₁₁H₁₇N₃O₄ MW: 255,3 g/mol Bod tání: ∼ 198 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00065576 CAS číslo: 17791-52-5 EINECS: 241-768-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...