Diosmin
Dodavatel: Thermo Scientific
Synonyms:
5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one
J62073.06
J62073.14
J62073.06EA
1640
CZK
J62073.06
J62073.14
Diosmin
Diosmin
CAS No.: 520-27-4
Molecular Formula: C28H32O15
Formula Weight: 608.50
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Pale yellow to brownish
Melting Point: 274°
Merck Reference: 14,3297
MDL No.: MFCD00009772
Natural flavanoid with anti-inflammatory effects
Vzorec:
C₂₈H₃₂O₁₅ MW: 608,5 g/mol Bod tání: 274 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009772 CAS číslo: 520-27-4 EINECS: 208-289-7 Merck Index: 14,3297 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...