Copper(I) bromide dimethyl sulphide complex, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
230502-10G
230502-50G
SIAL230502-10GEA
3130
CZK
SIAL230502-10G
SIAL230502-50G
Copper(I) bromide dimethyl sulphide complex, Sigma-Aldrich®
Copper(I) bromide dimethyl sulphide complex
Vzorec:
C₂H₆BrCuS MW: 205.58 g/mol |
MDL Number:
MFCD00043295 CAS číslo: 54678-23-8 EINECS: 259-292-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...