Tisk…

D-Glucoronic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: D-(+)Glucuronic acid, D-Glucuronic acid, D-(+)-glucuronic acid, (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid

M-Clarity™ quality level = MQ200

G5269-100G G5269-10G G5269-10MG G5269-25G G5269-50G
SIALG5269-100GEA 34270 CZK
SIALG5269-100G SIALG5269-10G SIALG5269-10MG SIALG5269-25G SIALG5269-50G
D-Glucoronic acid, Sigma-Aldrich®
D-Glucoronic acid
Vzorec: C₆H₁₀O₇
MW: 194.139 g/mol
MDL Number: MFCD00166981
CAS číslo: 6556-12-3
EINECS: 229-486-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR