Tisk…

Sodium-D(+)-glucuronate monohydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: D-Glucuronic acid sodium salt hydrate, D-Glucuronic acid sodium salt monohydrate, D(+)-Glucuronic acid, sodium salt, monohydrate, Sodium (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoate hydrate

M-Clarity™ quality level = MQ300

G8645-100G G8645-10G G8645-1KG G8645-25G G8645-50G G8645-5G
SIALG8645-100GEA 15940 CZK
SIALG8645-100G SIALG8645-10G SIALG8645-1KG SIALG8645-25G SIALG8645-50G SIALG8645-5G
Sodium-D(+)-glucuronate monohydrate, Sigma-Aldrich®
Sodium-D(+)-glucuronate monohydrate
Vzorec: C₆H₁₀O₇·1H₂O
MW: 234.14 g/mol (216.14 anhy)
MDL Number: MFCD00151051
CAS číslo: 207300-70-7
EINECS: 239-065-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR