Tisk…
Phenol (equilibrated) : Chloroform : iso-Amyl alcohol (25:24:1)% vol. stabilizovaný
Dodavatel: PanReac AppliChem
Nebezpečí
Stabilization: Stabilised with 8-hydroxyquinoline
A0889.0100 A0889.0500
A0889.0100EA 2030 CZK
A0889.0100 A0889.0500
Phenol (equilibrated) : Chloroform : iso-Amyl alcohol (25:24:1)% vol. stabilizovaný
Phenol (equilibrated) : Chloroform : iso-Amyl alcohol
Storage Temperature: Ambient
UN: 2810
ADR: 6.1,II