4-(1-Pyrrolidinyl)pyridine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Pyrrolidinopyridine
M-Clarity™ quality level = MQ200
83275-25G
83275-5G
SIAL83275-25GEA
6660
CZK
SIAL83275-25G
SIAL83275-5G
4-(1-Pyrrolidinyl)pyridine, Sigma-Aldrich®
4-(1-Pyrrolidinyl)pyridine
Vzorec:
C₉H₁₂N₂ MW: 148,21 g/mol |
MDL Number:
MFCD00006415 CAS číslo: 2456-81-7 EINECS: 219-534-2 UN: 2923 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...