Tisk…

Zinc (II) phthalocyanine (Sulphate ionophore II) ≥95%

Dodavatel: Thermo Fisher Scientific

Synonyms: Zinc phthalocyanine, 29H,31H-Phthalocyanine, zinc complex

39553.04 39553.09 39553.18
39553.04EA 1550 CZK
39553.04 39553.09 39553.18
Zinc (II) phthalocyanine (Sulphate ionophore II) ≥95%
Zinc (II) phthalocyanine (Sulphate ionophore II)

Purity: 95% CAS No.: 14320-04-8 Molecular Formula: C32H16N8Zn Formula Weight: 577.91 Physical Form: Powder Appearance: Purple Odour: Odourless MDL No.: MFCD00041955

Vzorec: C₃₂H₁₆N₈Zn
MW: 577.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041955
CAS číslo: 14320-04-8
EINECS: 238-262-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR