[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex
Dodavatel: abcr
Synonyms:
Dichloro 1,1'-bis(diphenylphosphino)ferrocene palladium (II) dichloromethane, [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) Dichloride Dichloromethane Adduct (1:1), Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), complex with dichloromethane, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane complex
AB125627.0001
AB125627.0005
ABCRAB125627.0001EA
0
CZK
ABCRAB125627.0001
ABCRAB125627.0005
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex
Vzorec:
C₃₅H₃₀Cl₄FeP₂Pd |
MDL Number:
MFCD00792899 CAS číslo: 95464-05-4 |