Tisk…

Boron trifluoride ethylamine complex, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

292249-100G 292249-500G
SIAL292249-100GEA 2490 CZK
SIAL292249-100G SIAL292249-500G
Boron trifluoride ethylamine complex, Sigma-Aldrich®
Boron trifluoride ethylamine complex
Vzorec: C₂H₇BF₃N
MDL Number: MFCD00144277
CAS číslo: 75-23-0
EINECS: 200-852-5
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR