Dysprosium(III) 2,4-pentanedionate hydrate ≥99.9% (REO, rare earth oxide basis)
Dodavatel: Thermo Fisher Scientific
13219.06
13219.14
13219.06EA
2590
CZK
13219.06
13219.14
Dysprosium(III) 2,4-pentanedionate hydrate ≥99.9% (REO, rare earth oxide basis)
Dysprosium(III) 2,4-pentanedionate hydrate
Purity: 99.9%
CAS No.: 14637-88-8
Molecular Formula: C15H21DyO6.xH2O
Formula Weight: 459.83(anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
MDL No.: MFCD00013489
Vzorec:
C₁₅H₂₁DyO₆· xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00013489 CAS číslo: 14637-88-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...