Tisk…

4-(N,N-Diphenylamino)phenylboronic acid pinacol ester 95%

Dodavatel: Thermo Fisher Scientific

H51946.03 H51946.06
H51946.03EA 2020 CZK
H51946.03 H51946.06
4-(N,N-Diphenylamino)phenylboronic acid pinacol ester 95%
4-(N,N-Diphenylamino)phenylboronic acid pinacol ester

Purity: 95% Molecular Formula: C24H26BNO2 Formula Weight: 371.29 MDL No.: MFCD13195770

Vzorec: C₂₄H₂₆BNO₂
Storage Temperature: Ambient
MDL Number: MFCD13195770
CAS číslo: 267221-88-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR