Diclofenac sodium salt, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium salt, GP-45840, [o-(2,6-dichloroanilino) phenyl]acetic acid sodium salt, 2-[(2,6-Dichlorophenyl)amino]-benzeneacetic acid monosodium salt, Sodium [o-[(2,6-dichlorophenyl) amino] phenyl] acetate
M-Clarity™ quality level = MQ100
D6899-25G
D6899-10G
D6899-100G
SIALD6899-25GEA
9270
CZK
SIALD6899-25G
SIALD6899-10G
SIALD6899-100G
Diclofenac sodium salt, Sigma-Aldrich®
Diclofenac sodium salt
Vzorec:
C₁₄H₁₀Cl₂NNaO₂ MW: 318,1 g/mol |
MDL Number:
MFCD00082251 CAS číslo: 15307-79-6 EINECS: 239-346-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...