Diclofenac sodium salt
Dodavatel: Cayman Chemical
Synonyms:
2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium salt, GP-45840, [o-(2,6-dichloroanilino) phenyl]acetic acid sodium salt, 2-[(2,6-Dichlorophenyl)amino]-benzeneacetic acid monosodium salt, Sodium [o-[(2,6-dichlorophenyl) amino] phenyl] acetate
70680-5
70680-25
70680-50
70680-100
CAYM70680-5EA
1290
CZK
CAYM70680-5
CAYM70680-25
CAYM70680-50
CAYM70680-100
Diclofenac sodium salt
Diclofenac sodium salt
Vzorec:
C₁₄H₁₀Cl₂NNaO₂ |
MDL Number:
MFCD00082251 CAS číslo: 15307-79-6 EINECS: 239-346-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...