Bis(2,2,2-trifluoroethyl)amine 95%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L16709.03
L16709.06
L16709.03EA
2380
CZK
L16709.03
L16709.06
Bis(2,2,2-trifluoroethyl)amine 95%
Bis(2,2,2-trifluoroethyl)amine
Purity: 96%
CAS No.: 407-01-2
Molecular Formula: (CF3CH2)2NH
Formula Weight: 181.08
Refractive Index: 1.3010
Boiling Point: 70-71°
UN No.: 2735
MDL No.: MFCD00042094
Vzorec:
C₄H₅F₆N Bod varu: 70…71 °C Hustota: 1,100 Storage Temperature: Ambient |
MDL Number:
MFCD00042094 CAS číslo: 407-01-2 EINECS: 206-978-7 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...