Tisk…

N-Methyliminodiacetic acid 99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2,2'-(Methylazanediyl)diacetic acid

L02589.06 L02589.14
L02589.06EA 1530 CZK
L02589.06 L02589.14
N-Methyliminodiacetic acid 99%
N-Methyliminodiacetic acid

Purity: 99% CAS No.: 4408-64-4 Molecular Formula: C5H9NO4 Formula Weight: 147.13 Melting Point: ca 220° dec. Beilstein Reference: 1765449

Vzorec: C₅H₉NO₄
MW: 147.13 g/mol
Bod tání: ∼ 220 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00004284
CAS číslo: 4408-64-4
EINECS: 224-557-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR