Tisk…

Glycine hydrochloride (H-Gly-OH.HCl), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: H-Gly-OH·HCl, Glycine hydrochloride

M-Clarity™ quality level = MQ200

G2879-100G G2879-1KG G2879-500G
SIALG2879-100GEA 3850 CZK
SIALG2879-100G SIALG2879-1KG SIALG2879-500G
Glycine hydrochloride (H-Gly-OH.HCl), Sigma-Aldrich®
Glycine hydrochloride (H-Gly-OH.HCl)
Vzorec: C₂H₆ClNO₂
MW: 111,53 g/mol
MDL Number: MFCD00012872
CAS číslo: 6000-43-7
EINECS: 227-841-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR