D-(-)-Diisopropyl tartrate ≥98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
(2S,3S)-(-)-Diisopropyl tartrate, (2S,3S)-Diisopropyl 2,3-dihydroxysuccinate, D-(-)-Tartaric acid diisopropyl ester
A17362.14
A17362.22
A17362.14EA
2370
CZK
A17362.14
A17362.22
D-(-)-Diisopropyl tartrate ≥98%
D-(-)-Diisopropyl tartrate
Purity: 98%
CAS No.: 62961-64-2
Molecular Formula: C10H18O6
Formula Weight: 234.25
Flash Point: 110°(230°F)
Refractive Index: 1.4400
Boiling Point: 150-151°/11mm
Beilstein Reference: 5265431
Optical Rotation: -16° (neat)
MDL No.: MFCD00008876
Vzorec:
C₁₀H₁₈O₆ Bod varu: 150…151 °C (11 mmHg) Hustota: 1,117 Bod vzplanutí: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008876 CAS číslo: 62961-64-2 EINECS: 263-771-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...