N-Phenylglycine ethyl ester ≥99%
Dodavatel: Thermo Scientific
Synonyms:
N-Phenylglycine Ethyl Ester, Phenylamino-acetic acid ethyl ester
B22282.09
B22282.18
B22282.09EA
814
CZK
B22282.09
B22282.18
N-Phenylglycine ethyl ester ≥99%
N-Phenylglycine ethyl ester
Purity: 99%
CAS No.: 2216-92-4
Molecular Formula: C6H5NHCH2CO2CH2CH3
Formula Weight: 179.22
Storage Temperature: 2°C to 8°C
Melting Point: 55-58°
MDL No.: MFCD00038315
Vzorec:
C₁₀H₁₃NO₂ MW: 179,22 g/mol Bod tání: 55…58 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00038315 CAS číslo: 2216-92-4 EINECS: 218-702-2 Merck Index: 14,7292 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...