Tisk…

N-Boc-glycine (boc-gly-OH), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Boc-Gly-OH

15420-100G 15420-25G 15420-5G
SIAL15420-100GEA 6950 CZK
SIAL15420-100G SIAL15420-25G SIAL15420-5G
N-Boc-glycine (boc-gly-OH), Sigma-Aldrich®
N-Boc-glycine (boc-gly-OH)
Vzorec: C₇H₁₃NO₄
MW: 175,2 g/mol
MDL Number: MFCD00002690
CAS číslo: 4530-20-5
EINECS: 224-864-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR