Tisk…

2-Ethylhexyl-4-(dimethylamino)benzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Ethylhexyl 4-(dimethylamino)benzoate

437395-100ML 437395-500ML
SIAL437395-100MLEA 1920 CZK
SIAL437395-100ML SIAL437395-500ML
2-Ethylhexyl-4-(dimethylamino)benzoate, Sigma-Aldrich®
2-Ethylhexyl-4-(dimethylamino)benzoate
Vzorec: C₁₇H₂₇NO₂
MW: 277,40 g/mol
MDL Number: MFCD00017526
CAS číslo: 21245-02-3
EINECS: 244-289-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR