Cyclopentadienyltitanium trichloride, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
234834-1G
234834-5G
SIAL234834-1GEA
1670
CZK
SIAL234834-1G
SIAL234834-5G
Cyclopentadienyltitanium trichloride, Sigma-Aldrich®
Cyclopentadienyltitanium trichloride
Vzorec:
C₅Cl₃Ti |
MDL Number:
MFCD00003724 CAS číslo: 1270-98-0 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...