Tisk…
Chemical resistant gloves, uvex profapren CF33
Dodavatel: UVEX
6011904 6011903 6011902 6011901 6011905
UVEX6011904EA 2300 CZK
UVEX6011904 UVEX6011903 UVEX6011902 UVEX6011901 UVEX6011905
Chemical resistant gloves, uvex profapren CF33
Rukavice
These polychloroprene gloves with latex inside and cotton flock lining provide protection against a wide range of chemical and mechanical hazards.
  • Flexible, durable
  • Cotton flock lining makes the glove softer and more comfortable to wear
  • Lining helps absorb perspiration
  • Silicone-free
Ideal for use in the chemical and metal industry, for cleaning and for painting work.
Certifikace: EN ISO 374-1:2016 Type A (AKLMNO), EN 388:2016 (3131X).