Tisk…
Erlenmeyer flasks, graduated, with screw cap
Dodavatel: BELLCO GLASS
2514-00050 2514-03000 2514-00025 2514-02000
BELC2514-00050EA 37080 CZK
BELC2514-00050 BELC2514-03000 BELC2514-00025 BELC2514-02000
Erlenmeyer flasks, graduated, with screw cap
Baňky Baňky dle Erlenmeyera
Manufactured from type 1 class A borosilicate glass, these flasks are ideal for media storage and mixing. Long-skirted phenolic rubber lined screw cap protects flask contents from contamination.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Graduated to approximate volumes