Hydroxylapatite, fast flow
Dodavatel: Thermo Scientific
|
Varování
|
J60076.18
J60076.09
J60076.18EA
11020
CZK
J60076.18
J60076.09
Hydroxylapatite, fast flow
Hydroxylapatite
CAS No.: 1306-06-5
Molecular Formula: HCa5O13P3
Formula Weight: 502.32
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Merck Reference: 14,3467
MDL No.: MFCD00010904
Used for fractionation and purification of proteins, enzymes, plasmids and nucleic acids
Vzorec:
Ca₅HO₁₃P₃ Storage Temperature: Freezer |
MDL Number:
MFCD00010904 CAS číslo: 1306-06-5 EINECS: 215-145-7 Merck Index: 14,3467 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...