Tisk…
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) ≥98%
Dodavatel: Thermo Scientific
Synonyms: 5-Bromo-4-chloro-1H-indol-3-yl phosphate sodium, X-phosphate disodium salt, Sodium 5-bromo-4-chloro-1H-indol-3-yl phosphate
J64451.MD J64451.03
J64451.MDEA 3990 CZK
J64451.MD J64451.03
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) ≥98%
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
Vzorec: C₈H₄BrClNNa₂O₄P
MW: 370,43 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036757
CAS číslo: 102185-33-1