Tisk…

4-Chlorobenzo[b]thiophene-2-carboxylic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

H33015.03 H33015.06
H33015.03EA 1490 CZK
H33015.03 H33015.06
4-Chlorobenzo[b]thiophene-2-carboxylic acid ≥97%
4-Chlorobenzo[b]thiophene-2-carboxylic acid

Purity: 97% CAS No.: 23967-57-9 Molecular Formula: C9H5ClO2S Formula Weight: 212.65 Melting Point: ca 233° subl. MDL No.: MFCD06655155

Vzorec: C₉H₅ClO₂S
Bod tání: ∼ 233 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD06655155
CAS číslo: 23967-57-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR