Tisk…

Methyl green zinc chloride salt

Dodavatel: Thermo Scientific
Varování

J61509.14 J61509.22
J61509.14EA 3470 CZK
J61509.14 J61509.22
Methyl green zinc chloride salt
Methyl green zinc chloride salt

CAS No.: 7114-03-6 Molecular Formula: C27H35Cl2N3.ZnCl2 Formula Weight: 653.24 Physical Form: Powder Appearance: Dark green Melting Point: 300° Beilstein Reference: 3914106 MDL No.: MFCD00151094


DNA stain useful as tracking dye in acid buffers for electrophoresis. Binds selectively to AT-rich regions of native DNA.

Vzorec: C₂₇H₃₅BrClN₃·xZnCl₂
MW: 653,24 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00151094
CAS číslo: 7114-03-6
EINECS: 230-415-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR