Tisk…

Carbenicillin disodium salt

Dodavatel: Thermo Fisher Scientific
Nebezpečí

J61949.03 J61949.06 J61949.14
J61949.03EA 2880 CZK
J61949.03 J61949.06 J61949.14
Carbenicillin disodium salt
Carbenicillin disodium salt

CAS No.: 4800-94-6 Molecular Formula: C17H16N2Na2O6S Formula Weight: 422.36 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white Beilstein Reference: 5722128 Merck Reference: 14,1792 MDL No.: MFCD00077683


Water-soluble antibiotic effective against Gram-negative bacteria

Vzorec: C₁₇H₁₆N₂O₆S·2Na
Storage Temperature: Refrigerator
MDL Number: MFCD00077683
CAS číslo: 4800-94-6
EINECS: 225-360-8
Merck Index: 14,1792

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR