Tisk…

Spermine tetrahydrochloride 99%

Dodavatel: Thermo Scientific
Varování

Synonyms: N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine) tetrahydrochloride

J63060.03 J63060.06 J63060.14
J63060.03EA 1270 CZK
J63060.03 J63060.06 J63060.14
Spermine tetrahydrochloride 99%
Spermine tetrahydrochloride

Purity: 99% CAS No.: 306-67-2 Molecular Formula: C10H26N4.4HCl Formula Weight: 348.19 Physical Form: Powder Appearance: White to off-white Beilstein Reference: 3911771 MDL No.: MFCD00012914


Binds to the polyamine modulatory site of NMDA

Vzorec: C₁₀H₃₀Cl₄N₄
MW: 348,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012914
CAS číslo: 306-67-2
EINECS: 206-189-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR