Tisk…

5-Fluorouridine ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

J62083.03 J62083.06
J62083.03EA 1710 CZK
J62083.03 J62083.06
5-Fluorouridine ≥97%
5-Fluorouridine

Purity: 97% CAS No.: 316-46-1 Molecular Formula: C9H11FN2O6 Formula Weight: 262.19 Physical Form: Powder Appearance: White Beilstein Reference: 33662 MDL No.: MFCD00036832

Vzorec: C₉H₁₁FN₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00036832
CAS číslo: 316-46-1
EINECS: 206-260-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR