Tisk…
Kanamycin sulphate cell culture reagent
Dodavatel: Thermo Fisher Scientific
Nebezpečí
Synonyms: Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate
J60668.03 J60668.06
J60668.03EA 1900 CZK
J60668.03 J60668.06
Kanamycin sulphate cell culture reagent
Kanamycin sulphate
CAS No.: 25389-94-0 Molecular Formula: C18H36N4O11.H2SO4 Formula Weight: 582.58 Physical Form: Powder Appearance: White to off-white Solubility: Soluble in water at 10mg/ml MDL No.: MFCD00070253
Vzorec: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070253
CAS číslo: 25389-94-0
EINECS: 246-933-9
Merck Index: 14,5281