Tisk…
Dextran, M.W. ~ 6,000
Dodavatel: Thermo Fisher Scientific
J62775.09 J62775.18 J62775.22 J62775.36
J62775.09EA 1300 CZK
J62775.09 J62775.18 J62775.22 J62775.36
Dextran, M.W. ~ 6,000
Dextran
CAS No.: 9004-54-0 Molecular Formula: (C6H10O5)n Physical Form: Powder Appearance: White to off-white Merck Reference: 14,2948 MDL No.: MFCD00130935
Vzorec: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00130935
CAS číslo: 9004-54-0
EINECS: 232-677-5
Merck Index: 14,2948