Tisk…

4-Aminopyridine ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Pyridin-4-amine, 4-Pyridylamine

J61470.14 J61470.22
J61470.14EA 2080 CZK
J61470.14 J61470.22
4-Aminopyridine ≥99%
4-Aminopyridine

Purity: 99+% CAS No.: 504-24-5 Molecular Formula: C5H6N2 Formula Weight: 94.12 Flash Point: 164°(327°F) Physical Form: Crystalline powder Density: 1.26 Appearance: White Odour: Unpleasant Melting Point: 157-161° Boiling Point: 273-274° Beilstein Reference: 105782 Merck Reference: 14,3933 UN No.: 2671 MDL No.: MFCD00006439


A potassium channel blocker

Vzorec: C₅H₆N₂
MW: 94,12 g/mol
Bod varu: 273…274 °C
Bod tání: 157…161 °C
Hustota: 1,26
Bod vzplanutí: 164 °C (327 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006439
CAS číslo: 504-24-5
EINECS: 207-987-9
UN: 2671
ADR: 6.1,II
Merck Index: 14,3933

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR