Fenol 99% unstabilised, Ultrapure
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Phenylalcohol, Carbolic acid, Hydroxybenzene
J64011.22
J64011.36
J64011.22EA
1540
CZK
J64011.22
J64011.36
Fenol 99% unstabilised, Ultrapure
Fenol
Purity: 99%
CAS No.: 108-95-2
Molecular Formula: C6H5OH
Formula Weight: 94.11
Flash Point: 79°(175°F)
Physical Form: Crystalline
Density: 1.071
Sensitivity: Air Sensitive
Appearance: White
Odour: Sweet
Melting Point: 40-43°
Boiling Point: 181°
Beilstein Reference: 969616
Merck Reference: 14,7241
UN No.: 1671
MDL No.: MFCD00002143
Vzorec:
C₆H₅OH MW: 94.11 Bod varu: 181 °C Bod tání: 40…43 °C Hustota: 1.071 Bod vzplanutí: 79 °C (175 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002143 CAS číslo: 108-95-2 EINECS: 203-632-7 UN: 1671 ADR: 6.1 II REACH: 01-2119471329-32 Merck Index: 14,7241 |