Mycophenolic acid ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
J61905.03
J61905.06
J61905.09
J61905.03EA
1400
CZK
J61905.03
J61905.06
J61905.09
Mycophenolic acid ≥98%
Mycophenolic acid
Purity: 98%
CAS No.: 24280-93-1
Molecular Formula: C17H20O6
Formula Weight: 320.34
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Melting Point: 139-141°
Solubility: Soluble in methanol at 50mg/ml
Beilstein Reference: 1295848
Merck Reference: 14,6327
MDL No.: MFCD00036814
Immunosupressant agent. Blocks inosine monophosphate dehydrogenase in the guanosine monophosphate pathway
Vzorec:
C₁₇H₂₀O₆ Bod tání: 139…141 °C Storage Temperature: Freezer |
MDL Number:
MFCD00036814 CAS číslo: 24280-93-1 EINECS: 246-119-3 UN: 3077 ADR: 9,III Merck Index: 14,6327 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...