Daidzein ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Daidzeol, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',7-Dihydroxyisoflavone, 7-Hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
J63763.ME
J63763.03
J63763.MC
J63763.MEEA
3850
CZK
J63763.ME
J63763.03
J63763.MC
Daidzein ≥98%
Daidzein
Purity: 98+%
CAS No.: 486-66-8
Molecular Formula: C15H10O4
Formula Weight: 254.24
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Melting Point: 328-332° dec.
Beilstein Reference: 231523
Merck Reference: 14,2801
MDL No.: MFCD00016954
Analog of genistein and an agonist at estrogen recpetors
Vzorec:
C₁₅H₁₀O₄ MW: 254,24 g/mol Bod varu: 513 °C (1013 hPa) Bod tání: 328…332 °C (rozkl.) Storage Temperature: Freezer |
MDL Number:
MFCD00016954 CAS číslo: 486-66-8 EINECS: 207-635-4 Merck Index: 14,2801 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...