Tisk…

Eskulin seskihydrát

Dodavatel: Thermo Fisher Scientific

Synonyms: 6,7-Dihydroxycoumarin 6-glucoside sesquihydrate, Esculin sesquihydrate

J63114.06 J63114.14
J63114.06EA 1530 CZK
J63114.06 J63114.14
Eskulin seskihydrát
Eskulin seskihydrát

CAS No.: 66778-17-4 Molecular Formula: C15H16O9.1.5H2O Formula Weight: 367.31 (340.29anhy) Physical Form: Powder Appearance: White to off-white Melting Point: 202-210° MDL No.: MFCD00149492


A naturally occuring antioxidant

Vzorec: C₁₅H₁₆O₉·1,5H₂O
MW: 367.31
Bod tání: 202…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00149492
CAS číslo: 66778-17-4
EINECS: 208-517-5
Merck Index: 14,3698

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR