Tisk…

Polymyxin B sulphate

Dodavatel: Thermo Fisher Scientific
Varování

J63074.03 J63074.06
J63074.03EA 4230 CZK
J63074.03 J63074.06
Polymyxin B sulphate
Polymyxin B sulphate

Antibiotic for Gram-negative bacteria.


  • White to pale yellow powder


Soluble in water at 50mg/ml

Vzorec: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00131991
CAS číslo: 1405-20-5
EINECS: 215-774-7
Merck Index: 14,7573

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR