Oleyl alcohol ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
cis-Octadec-9-en-1-ol, cis-9-Octadecen-1-ol
H33475.03
H33475.14
H33475.06
H33475.03EA
4840
CZK
H33475.03
H33475.14
H33475.06
Oleyl alcohol ≥99%
Oleyl alcohol
Purity: 99+%
CAS No.: 143-28-2
Molecular Formula: C18H36O
Formula Weight: 268.48
Storage Temperature: -30°C to -10°C
Flash Point: 180°(356°F)
Refractive Index: 1.4600
Melting Point: ca 6°
Boiling Point: 206-207/13mm
Beilstein Reference: 1723962
MDL No.: MFCD00002993
Vzorec:
C₁₈H₃₆O MW: 268,48 g/mol Bod varu: 206…207 °C (13 mmHg) Bod tání: ∼ 6 °C Hustota: 0,849 Bod vzplanutí: 180 °C (356 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00002993 CAS číslo: 143-28-2 EINECS: 205-597-3 Merck Index: 12,06968 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...