2-Amino-2-methyl-1,3-propanediol ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
J63144.14
J63144.22
J63144.30
J63144.14EA
934
CZK
J63144.14
J63144.22
J63144.30
2-Amino-2-methyl-1,3-propanediol ≥99%
2-Amino-2-methyl-1,3-propanediol
Purity: 99+%
CAS No.: 115-69-5
Molecular Formula: C4H11NO2
Formula Weight: 105.14
Physical Form: Powder
Appearance: White
Melting Point: 100-110°
Boiling Point: 151°/10mm
Beilstein Reference: 635708
Merck Reference: 14,448
MDL No.: MFCD00004678
A useful biological buffer
Vzorec:
C₄H₁₁NO₂ MW: 105,14 g/mol Bod varu: 151 °C (10 mmHg) Bod tání: 100…110 °C Hustota: 1,211 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004678 CAS číslo: 115-69-5 EINECS: 204-100-7 Merck Index: 14,448 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...