Tisk…

Penicillin G sodium salt

Dodavatel: Thermo Fisher Scientific
Nebezpečí

J63032.06 J63032.14 J63032.22
J63032.06EA 1150 CZK
J63032.06 J63032.14 J63032.22
Penicillin G sodium salt
Penicillin G sodium salt

CAS No.: 69-57-8 Molecular Formula: C16H17N2NaO4S Formula Weight: 356.37 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Beilstein Reference: 3834217 Merck Reference: 14,7094 MDL No.: MFCD00069666


Inhibits bacterial cell wall synthesis

Vzorec: C₁₆H₁₇N₂NaO₄S
Bod tání: 255 °C
Hustota: 1,38 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00069666
CAS číslo: 69-57-8
EINECS: 200-710-2
Merck Index: 14,7094

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR