Draselná sůl penicilinu G
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
6-(2-Phenylacetamido)penicillanic acid potassium salt, Potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Penicillin G potassium, Potassium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Benzylpenicillin potassium salt
CAS No.: 113-98-4
Molecular Formula: C16H17KN2O4S
Formula Weight: 372.48
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 3832841
Merck Reference: 14,7094
MDL No.: MFCD00036193
Inhibits bacterial cell wall synthesis
Vzorec:
C₁₆H₁₇KN₂O₄S MW: 372,49 g/mol Bod tání: 214…217 °C Hustota: 1,359 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00036193 CAS číslo: 113-98-4 EINECS: 204-038-0 Merck Index: 14,7094 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...