L-Cystin cell culture reagent
Dodavatel: Thermo Fisher Scientific
Synonyms:
(R)-(-)-Cystine, (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid), (H-Cys-OH)2
J61651.09
J61651.A1
J61651.09EA
869
CZK
J61651.09
J61651.A1
L-Cystin cell culture reagent
L-Cystin
CAS No.: 56-89-3
Molecular Formula: C6H12N2O4S2
Formula Weight: 240.30
Physical Form: Powder
Density: 1.68
Appearance: White
Melting Point:
Vzorec:
HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH MW: 240.3 Bod varu: 428 Bod tání: >240 °C (rozkl.) Hustota: 1.68 Storage Temperature: Ambient |
MDL Number:
MFCD00064228 CAS číslo: 56-89-3 EINECS: 200-296-3 Merck Index: 14,2782 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...