Tisk…

myo-Inozitol cell culture reagent

Dodavatel: Thermo Scientific

Synonyms: i-Inositol, D-Myo-Inositol, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol, Inositol

J62886.18 J62886.22 J62886.30
J62886.18EA 2560 CZK
J62886.18 J62886.22 J62886.30
myo-Inozitol cell culture reagent
myo-Inozitol

CAS No.: 87-89-8 Molecular Formula: C6H12O6 Formula Weight: 180.16 Physical Form: Crystalline powder Appearance: White Odour: Odourless Beilstein Reference: 1907329 Merck Reference: 14,4978 MDL No.: MFCD00077932


A lipotropic agent

Vzorec: C₆H₁₂O₆
MW: 180,16 g/mol
Bod varu: 291 °C (1013 hPa)
Bod tání: 222…227 °C
Hustota: 1,752 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00077932
CAS číslo: 87-89-8
EINECS: 201-781-2
Merck Index: 14,4978

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR