L-Tyrosin cell culture reagent
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH
J63511.09
J63511.22
J63511.36
J63511.A1
J63511.09EA
906
CZK
J63511.09
J63511.22
J63511.36
J63511.A1
L-Tyrosin cell culture reagent
L-Tyrosin
CAS No.: 60-18-4
Molecular Formula: C9H11NO3
Formula Weight: 181.19
Physical Form: Powder
Appearance: White to pale-brown
Melting Point:
Vzorec:
C₉H₁₁NO₃ MW: 181,19 g/mol Bod varu: 351 °C (1013 hPa) Bod tání: >300 °C Hustota: 1,41 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002606 CAS číslo: 60-18-4 EINECS: 200-460-4 Merck Index: 14,9839 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...