Tisk…

L(+)-cystein cell culture reagent

Dodavatel: Thermo Scientific
Varování

Synonyms: H-Cys-OH, (R)-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid

J63745.14 J63745.22
J63745.14EA 1400 CZK
J63745.14 J63745.22
L(+)-cystein cell culture reagent
L(+)-cystein

CAS No.: 52-90-4 Molecular Formula: C3H7NO2S Formula Weight: 121.16 Physical Form: Powder Sensitivity: Air Sensitive Appearance: White Melting Point: ca 220° dec. Beilstein Reference: 1721408 Merck Reference: 14,2781 Optical Rotation: +7.6° (c=5 in 5N HCl) MDL No.: MFCD00064306

Vzorec: HSCH₂CH(NH₂)CO₂H
MW: 121,16 g/mol
Bod varu: 254 °C (1013 hPa)
Bod tání: ∼ 220 °C (rozkl.)
Hustota: 1,523 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064306
CAS číslo: 52-90-4
EINECS: 200-158-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR