Streptomycin sulphate cell culture reagent
Dodavatel: Thermo Fisher Scientific
|
Varování
|
J61299.18
J61299.22
J61299.18EA
2060
CZK
J61299.18
J61299.22
Streptomycin sulphate cell culture reagent
Streptomycin sulphate
CAS No.: 3810-74-0
Molecular Formula: C21H39N7O12 .1.5H2SO4
Formula Weight: 728.69
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in water
Beilstein Reference: 3894995
Merck Reference: 14,8826
MDL No.: MFCD00037023
Inhibits initiation and causes misreading of rRNA in protein synthesis
Vzorec:
C₂₁H₃₉N₇O₁₂·1.5H₂SO₄ Bod tání: 250 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00037023 CAS číslo: 3810-74-0 EINECS: 223-286-0 UN: 2811 ADR: 6.1,III Merck Index: 14,8826 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...